Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASPH SIRNA Assays ›
Gene Symbol:
Asph
Gene Name:
aspartate-beta-hydroxylase
Gene Aliases:
2310005F16Rik, 3110001L23Rik, AI848629, AW261690, AW561901, BAH, C79816, cI-37
Chromosome Location:
Chr.4: 9449085 - 9669344 on Build GRCm38
UniGene ID:
Mm.222206
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS250082
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Asph
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001177849.1
NP_001171320.1
3
550
NM_001177850.1
NP_001171321.1
3
550
NM_001177851.1
NP_001171322.1
3
550
NM_001177852.1
NP_001171323.1
3
351
NM_001177853.1
NP_001171324.1
3
351
NM_001177854.1
NP_001171325.1
3
351
NM_001177855.1
NP_001171326.1
3
550
NM_001177856.1
NP_001171327.1
4
595
NM_001290367.1
NP_001277296.1
3
357
NM_023066.3
NP_075553.2
3
550
NM_133723.2
NP_598484.2
3
351
GenBank mRNA
AF221854.1
-
3
326
AF223413.1
-
3
351
AF223414.1
-
3
361
AF289486.1
-
3
472
AF289487.1
-
3
472
AF289488.1
-
3
472
AF289490.1
-
3
308
AF289491.1
-
3
308
AF289492.1
-
3
308
AF302653.2
-
3
250
AF302654.1
-
3
303
AF302655.1
-
3
230
AK009170.1
-
3
276
AK009903.1
-
3
287
AK014674.1
-
3
284
AK017453.1
-
3
283
AK028679.1
-
3
282
AK030293.1
-
3
474
AK030694.1
-
3
285
AK032420.1
-
4
518
AK035735.1
-
3
359
AK049506.1
-
3
474
AK151787.1
-
3
473
AK160984.1
-
3
475
AK165260.1
-
3
496
BC015281.1
-
3
459
BC061098.1
-
4
541
BC128299.1
-
3
279
BC152365.1
-
3
279
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
pattern specification process
negative regulation of cell proliferation
positive regulation of calcium ion transport into cytosol
peptidyl-amino acid modification
regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
regulation of protein stability
activation of store-operated calcium channel activity
response to ATP
limb morphogenesis
peptidyl-aspartic acid hydroxylation
positive regulation of proteolysis
positive regulation of transcription, DNA-templated
regulation of cytosolic calcium ion concentration
oxidation-reduction process
palate development
face morphogenesis
calcium ion transmembrane transport
cellular response to calcium ion
positive regulation of intracellular protein transport
activation of cysteine-type endopeptidase activity
regulation of protein depolymerization
peptide-aspartate beta-dioxygenase activity
calcium ion binding
protein binding
oxidoreductase activity
metal ion binding
dioxygenase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0