Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JMJD6 SIRNA Assays ›
          Gene Symbol:
          Jmjd6
          Gene Name:
          jumonji domain containing 6
          Gene Aliases:
          5730436I23Rik, D11Ertd195e, PSR, PtdSerR, Ptdsr, mKIAA0585
          Chromosome Location:
          Chr.11: 116837432 - 116843449 on Build GRCm38
          UniGene ID:
          Mm.481684
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS272577
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Jmjd6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033398.2 NP_203971.2 3 951
          GenBank mRNA AF304118.1 - 3 884
          AK122317.1 - 3 792
          BC006067.1 - 3 760
          BC011503.1 - 3 783
          BC056629.1 - 3 951 5033487
          BC117737.1 - 2 592
          CX210791.1 - 3 657
          EF527404.1 - 3 951
          EF527405.1 - 3 737

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001012143, XM_006247806, BC079012.

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          kidney development
          sprouting angiogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          cell surface receptor signaling pathway
          multicellular organism development
          heart development
          RNA splicing
          chromatin modification
          peptidyl-lysine hydroxylation to 5-hydroxy-L-lysine
          cell differentiation
          lung development
          T cell differentiation in thymus
          macrophage activation
          apoptotic cell clearance
          recognition of apoptotic cell
          regulation of mRNA splicing, via spliceosome
          erythrocyte development
          oxidation-reduction process
          retina development in camera-type eye
          histone H3-R2 demethylation
          histone H4-R3 demethylation
          sulfonate dioxygenase activity
          RNA binding
          single-stranded RNA binding
          receptor activity
          iron ion binding
          protein binding
          oxidoreductase activity
          2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
          procollagen-proline dioxygenase activity
          histone demethylase activity (H3-R2 specific)
          histone demethylase activity (H4-R3 specific)
          hypophosphite dioxygenase activity
          identical protein binding
          DNA-N1-methyladenine dioxygenase activity
          metal ion binding
          dioxygenase activity
          C-19 gibberellin 2-beta-dioxygenase activity
          C-20 gibberellin 2-beta-dioxygenase activity
          peptidyl-lysine 5-dioxygenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0