Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTG1 SIRNA Assays ›
          Gene Symbol:
          Actg1 Actb Lrrc58
          Gene Name:
          actin, gamma, cytoplasmic 1
          actin, beta
          leucine rich repeat containing 58
          Gene Aliases:
          AL023024, Actg, Actl, E51
          Actx, E430023M04Rik, beta-actin
          1810012N18Rik, C330018J07Rik
          Chromosome Location:
          Chr.11: 120345689 - 120348491 on Build GRCm38
          Chromosome Location:
          Chr.5: 142903116 - 142906724 on Build GRCm38
          Chromosome Location:
          Chr.16: 37868400 - 37888857 on Build GRCm38
          UniGene ID:
          Mm.196173
          Mm.328431
          Mm.390882
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID MSS272991
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Actg1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009609.2 NP_033739.1 4 626
          GenBank mRNA AF195094.1 - 4 656
          AK012251.1 - 4 638
          AK076081.1 - 4 635
          AK087983.1 - 4 635
          AK149927.1 - 4 632
          AK150209.1 - 4 632
          AK152202.1 - 4 639
          AK153446.1 - 4 637
          AK157726.1 - 4 635
          AK159867.1 - 4 638
          AK162157.1 - 4 632
          AK183894.1 - 1 14
          AK208587.1 - 1 14
          AK208588.1 - 1 14
          AK209715.1 - 1 14
          BC003337.1 - 4 621 3257312
          BC018571.1 - 4 621
          BC021796.1 - 4 623 4218945
          BC023248.1 - 4 626 3499390
          BC099371.1 - 4 618 30841085
          M21495.1 - 4 599
          X13055.1 - 3 541
          Gene Symbol: Actb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK075973.1 - 4 639
          AK078935.1 - 4 641
          AK088691.1 - 4 638
          AK145191.1 - 4 641
          AK145196.1 - 4 641
          AK145308.1 - 4 641
          AK147787.1 - 4 638
          AK149920.1 - 4 641
          AK150545.1 - 4 642
          AK150553.1 - 4 642
          AK150558.1 - 4 642
          AK150570.1 - 4 643
          AK150662.1 - 4 640
          AK150675.1 - 4 640
          AK150711.1 - 4 641
          AK150854.1 - 4 642
          AK150866.1 - 4 641
          AK150879.1 - 4 641
          AK151010.1 - 4 638
          AK151136.1 - 4 641
          AK151145.1 - 4 638
          AK151159.1 - 4 641
          AK151166.1 - 4 638
          AK151190.1 - 4 641
          AK151202.1 - 4 641
          AK151226.1 - 4 638
          AK151277.1 - 4 638
          AK151350.1 - 4 638
          AK151385.1 - 4 641
          AK151386.1 - 4 638
          AK151398.1 - 4 641
          AK151468.1 - 4 638
          AK151712.1 - 4 643
          AK151898.1 - 4 639
          AK151956.1 - 4 638
          AK151995.1 - 4 641
          AK151999.1 - 4 641
          AK152043.1 - 4 642
          AK152052.1 - 4 639
          AK152083.1 - 4 642
          AK152218.1 - 4 642
          AK152433.1 - 4 639
          AK152470.1 - 4 639
          AK152591.1 - 4 642
          AK152615.1 - 4 641
          AK152651.1 - 4 641
          AK152671.1 - 4 642
          AK152813.1 - 4 642
          AK152841.1 - 4 642
          AK152844.1 - 4 638
          AK153461.1 - 4 638
          AK159759.1 - 4 641
          AK159834.1 - 4 638
          AK160029.1 - 4 638
          AK166349.1 - 4 641
          AK166498.1 - 4 641
          AK167117.1 - 4 641
          AK167825.1 - 4 641
          AK167960.1 - 4 641
          AK187650.1 - 1 53
          AK203791.1 - 1 14
          AK204356.1 - 1 75
          BC138611.1 - 4 606 8861631
          BC138614.1 - 4 606 8861634
          J04181.1 - 4 622
          X03672.1 - 4 640
          Gene Symbol: Lrrc58
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_177093.3 NP_796067.2 4 2807
          GenBank mRNA AK147818.1 - 4 2783

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001127449, NM_031144, XM_002729196, XM_002728532, FQ231596, and others.

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          sarcomere organization
          platelet aggregation
          biological_process
          nucleotide binding
          structural constituent of cytoskeleton
          protein binding
          ATP binding
          ubiquitin protein ligase binding
          identical protein binding
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0