Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGER SIRNA Assays ›
          Gene Symbol:
          Ager
          Gene Name:
          advanced glycosylation end product-specific receptor
          Gene Aliases:
          RAGE
          Chromosome Location:
          Chr.17: 34597410 - 34600937 on Build GRCm38
          UniGene ID:
          Mm.3383
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS273058
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ager
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271422.1 NP_001258351.1 7 759
          NM_001271423.1 NP_001258352.1 7 732
          NM_001271424.1 NP_001258353.1 6 854
          NM_007425.3 NP_031451.2 7 759
          GenBank mRNA AB207883.1 - 7 722
          AK004802.1 - 7 749
          BC061182.1 - 7 747 30300427
          EU520325.1 - 7 715
          EU570240.1 - 7 715
          EU570241.1 - 6 910
          EU570242.1 - 7 715
          EU570243.1 - 7 715
          EU570244.1 - 6 910
          EU570245.1 - 6 910
          EU570246.1 - 6 872
          EU570247.1 - 6 910
          EU906863.1 - 7 688
          EU906864.1 - 6 910
          GU164727.1 - 6 829
          GU164728.1 - 7 734
          GU164730.1 - 7 734
          GU164731.1 - 7 737
          KC692918.1 - 7 715
          L33412.1 - 7 723

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053336, XM_006256025, BC084697, GU164715, GU164720, and others.

          Panther Classification:

          Molecular Function -

          cell adhesion molecule immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          negative regulation of endothelial cell proliferation
          inflammatory response
          negative regulation of cell adhesion
          JAK-STAT cascade
          positive regulation of autophagy
          negative regulation of endothelial cell migration
          positive regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          positive regulation of fibroblast migration
          positive regulation of smooth muscle cell migration
          positive regulation of cell migration
          neuron projection development
          negative regulation of collagen biosynthetic process
          positive regulation of apoptotic process
          positive regulation of JUN kinase activity
          positive regulation of neuron apoptotic process
          positive regulation of fibroblast proliferation
          positive regulation of smooth muscle cell proliferation
          regulation of inflammatory response
          positive regulation of inflammatory response
          induction of positive chemotaxis
          positive regulation of NF-kappaB transcription factor activity
          regulation of DNA binding
          calcium ion homeostasis
          positive regulation of phagocytosis, engulfment
          protein localization to membrane
          positive regulation of potassium ion transmembrane transporter activity
          positive regulation of neuron death
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          positive regulation of type B pancreatic cell apoptotic process
          protein binding
          identical protein binding
          S100 protein binding
          advanced glycation end-product receptor activity
          high mobility group box 1 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0