Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BAX SIRNA Assays ›
          Gene Symbol:
          Bax
          Gene Name:
          BCL2-associated X protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 45461695 - 45466903 on Build GRCm38
          UniGene ID:
          Mm.19904
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS273362
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bax
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007527.3 NP_031553.1 5 569
          GenBank mRNA AK013298.1 - 5 479
          AK149994.1 - 5 462
          AK159233.1 - 5 534
          AY095934.1 - 4 916
          BC018228.1 - 5 514 3968903
          BC053380.1 - 5 486 6489475
          L22472.1 - 5 446

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017059, AF235993, CB586216, U49729, U59184.

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          neuron migration
          leukocyte homeostasis
          T cell homeostatic proliferation
          B cell homeostasis
          B cell apoptotic process
          kidney development
          release of cytochrome c from mitochondria
          protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          blood vessel remodeling
          myeloid cell homeostasis
          B cell negative selection
          B cell homeostatic proliferation
          positive regulation of B cell apoptotic process
          glycosphingolipid metabolic process
          regulation of nitrogen utilization
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          obsolete transformed cell apoptotic process
          cellular response to DNA damage stimulus
          inner mitochondrial membrane organization
          outer mitochondrial membrane organization
          germ cell development
          spermatogenesis
          nervous system development
          sex differentiation
          mitochondrial fusion
          cell proliferation
          negative regulation of cell proliferation
          male gonad development
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to DNA damage
          activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
          apoptotic mitochondrial changes
          fertilization
          response to wounding
          response to toxic substance
          response to salt stress
          post-embryonic development
          response to ionizing radiation
          establishment or maintenance of transmembrane electrochemical gradient
          response to gamma radiation
          positive regulation of calcium ion transport into cytosol
          hypothalamus development
          cerebral cortex development
          negative regulation of protein binding
          positive regulation of protein oligomerization
          negative regulation of endoplasmic reticulum calcium ion concentration
          release of matrix enzymes from mitochondria
          negative regulation of peptidyl-serine phosphorylation
          regulation of mammary gland epithelial cell proliferation
          cellular response to UV
          limb morphogenesis
          ectopic germ cell programmed cell death
          odontogenesis of dentin-containing tooth
          regulation of apoptotic process
          positive regulation of apoptotic process
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of protein homodimerization activity
          regulation of protein heterodimerization activity
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          mitochondrial fragmentation involved in apoptotic process
          development of secondary sexual characteristics
          cellular respiration
          retinal cell programmed cell death
          positive regulation of developmental pigmentation
          negative regulation of fibroblast proliferation
          spermatid differentiation
          post-embryonic camera-type eye morphogenesis
          response to axon injury
          homeostasis of number of cells
          homeostasis of number of cells within a tissue
          protein oligomerization
          protein homooligomerization
          positive regulation of release of sequestered calcium ion into cytosol
          neuron apoptotic process
          regulation of cell cycle
          regulation of mitochondrial membrane potential
          transmembrane transport
          Sertoli cell proliferation
          retina development in camera-type eye
          positive regulation of apoptotic process involved in mammary gland involution
          vagina development
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          thymocyte apoptotic process
          mitochondrion morphogenesis
          cellular response to organic substance
          intrinsic apoptotic signaling pathway by p53 class mediator
          positive regulation of release of cytochrome c from mitochondria
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          intrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          positive regulation of endoplasmic reticulum unfolded protein response
          regulation of mitochondrial membrane permeability involved in apoptotic process
          positive regulation of mitochondrial membrane permeability involved in apoptotic process
          apoptotic process involved in patterning of blood vessels
          apoptotic process involved in embryonic digit morphogenesis
          regulation of mitochondrial membrane permeability involved in programmed necrotic cell death
          positive regulation of apoptotic DNA fragmentation
          retinal cell apoptotic process
          B cell receptor apoptotic signaling pathway
          negative regulation of apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of intrinsic apoptotic signaling pathway
          protein binding
          lipid binding
          channel activity
          heat shock protein binding
          protein complex binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          chaperone binding
          BH domain binding
          BH3 domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0