Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CALM1 SIRNA Assays ›
          Gene Symbol:
          Calm1
          Gene Name:
          calmodulin 1
          Gene Aliases:
          AI256814, AI327027, AI461935, AL024000, CaM, Calm
          Chromosome Location:
          Chr.12: 100199541 - 100209806 on Build GRCm38
          UniGene ID:
          Mm.285993
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS273469
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Calm1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009790.4 NP_033920.1 2, 3 217
          GenBank mRNA AK004673.1 - 2, 3 231
          AK013695.1 - 2, 3 226
          AK088141.1 - 2, 3 227
          AK145841.1 - 2, 3 204
          AK150288.1 - 2, 3 227
          AK150978.1 - 2, 3 232
          AK151001.1 - 2, 3 227
          AK151552.1 - 2, 3 227
          AK151784.1 - 2, 3 232
          AK151923.1 - 2, 3 230
          AK151992.1 - 2, 3 232
          AK152148.1 - 2, 3 230
          AK152715.1 - 2, 3 226
          AK152719.1 - 2, 3 227
          AK152850.1 - 2, 3 230
          AK152897.1 - 2, 3 227
          AK153004.1 - 2, 3 226
          AK153348.1 - 2, 3 230
          AK153426.1 - 2, 3 230
          AK153546.1 - 2, 3 230
          AK159762.1 - 2, 3 233
          AK160057.1 - 2, 3 233
          AK160508.1 - 2, 3 232
          AK161302.1 - 2, 3 188
          AK162314.1 - 2, 3 234
          AK166308.1 - 2, 3 227
          AK167353.1 - 2, 3 232
          AK168002.1 - 2, 3 230
          AK168241.1 - 2, 3 186
          AK168663.1 - 2, 3 226
          AK168803.1 - 2, 3 230
          AK169027.1 - 2, 3 232
          AK169055.1 - 2, 3 230
          AK169640.1 - 2, 3 232
          BC054805.1 - 2, 3 217 6838207
          M19381.1 - 2, 3 230
          X61432.1 - 2, 3 70

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031969, AF178845, FQ226971, FQ211760, FQ213227, and others.

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          positive regulation of DNA binding
          regulation of catalytic activity
          calcium ion binding
          adenylate cyclase binding
          protein domain specific binding
          enzyme regulator activity
          nitric-oxide synthase regulator activity
          type 3 metabotropic glutamate receptor binding
          phosphatidylinositol 3-kinase binding
          ion channel binding
          protein N-terminus binding
          calcium-dependent protein binding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0