Search
Search

| Gene Symbol: Drd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032054.1 | - | 1 | 134 | ||
| AK045456.1 | - | 1 | 323 | |||
| BC138059.1 | - | 1 | 191 | 8861078 | ||
| BC138060.1 | - | 1 | 191 | 8861079 | ||
| Gene Symbol: Gm26560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044390.1 | - | 1 | 1067 | ||