Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IFNG SIRNA Assays ›
          Gene Symbol:
          Ifng
          Gene Name:
          interferon gamma
          Gene Aliases:
          IFN-g, Ifg
          Chromosome Location:
          Chr.10: 118441047 - 118445892 on Build GRCm38
          UniGene ID:
          Mm.240327
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS275024
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ifng
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008337.3 NP_032363.1 1 188
          GenBank mRNA AK089574.1 - 1 189
          AY423847.1 - 1 37
          BC119063.1 - 1 117 8733812
          BC119065.1 - 1 117 8733814
          EF423643.1 - 1 80
          FJ617514.1 - 1 146
          FJ617515.1 - 1 146
          FJ617516.1 - 1 146
          K00083.1 - 1 149
          M28621.1 - 1 190

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine interferon superfamily intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          negative regulation of transcription from RNA polymerase II promoter
          neutrophil apoptotic process
          regulation of the force of heart contraction
          adaptive immune response
          CD8-positive, alpha-beta T cell differentiation involved in immune response
          regulation of transcription, DNA-templated
          apoptotic process
          inflammatory cell apoptotic process
          immune response
          humoral immune response
          cell cycle arrest
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to virus
          positive regulation of epithelial cell migration
          antigen processing and presentation
          neutrophil chemotaxis
          negative regulation of epithelial cell differentiation
          endoplasmic reticulum unfolded protein response
          negative regulation of myelination
          positive regulation of synaptic transmission, cholinergic
          negative regulation of interleukin-17 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-23 production
          positive regulation of tumor necrosis factor production
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          positive regulation of smooth muscle cell apoptotic process
          regulation of growth
          positive regulation of T cell proliferation
          positive regulation of tyrosine phosphorylation of Stat1 protein
          defense response to bacterium
          defense response to protozoan
          negative regulation of growth of symbiont in host
          negative regulation of growth of symbiont involved in interaction with host
          positive regulation of chemokine biosynthetic process
          positive regulation of interleukin-12 biosynthetic process
          positive regulation of MHC class II biosynthetic process
          positive regulation of interleukin-6 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of neuron differentiation
          positive regulation of osteoclast differentiation
          positive regulation of cell adhesion
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of isotype switching to IgG isotypes
          negative regulation of smooth muscle cell proliferation
          positive regulation of interleukin-1 beta secretion
          regulation of immune response
          regulation of insulin secretion
          sensory perception of mechanical stimulus
          positive regulation of membrane protein ectodomain proteolysis
          defense response to virus
          positive regulation of killing of cells of other organism
          positive regulation of fructose 1,6-bisphosphate 1-phosphatase activity
          positive regulation of fructose 1,6-bisphosphate metabolic process
          positive regulation of vitamin D biosynthetic process
          positive regulation of calcidiol 1-monooxygenase activity
          cellular response to lipopolysaccharide
          cellular response to interleukin-18
          extrinsic apoptotic signaling pathway
          regulation of neuronal action potential
          positive regulation of tumor necrosis factor (ligand) superfamily member 11 production
          regulation of hepatocyte proliferation
          cytokine activity
          interferon-gamma receptor binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0