Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL4 SIRNA Assays ›
          Gene Symbol:
          Il4
          Gene Name:
          interleukin 4
          Gene Aliases:
          BSF-1, Il-4
          Chromosome Location:
          Chr.11: 53612460 - 53618665 on Build GRCm38
          UniGene ID:
          Mm.276360
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS275079
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Il4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021283.2 NP_067258.1 4 458
          GenBank mRNA AB174765.1 - 4 437
          AF352783.1 - 4 439
          BC027514.1 - 4 437 3376587
          BX529138.1 - 3 486
          M13238.1 - 4 454
          M25892.1 - 4 486
          X03532.1 - 4 460

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          positive regulation of protein phosphorylation
          innate immune response in mucosa
          positive regulation of defense response to virus by host
          T-helper 1 cell lineage commitment
          positive regulation of immunoglobulin production
          negative regulation of acute inflammatory response
          negative regulation of chronic inflammatory response
          immune response
          cholesterol metabolic process
          positive regulation of cell proliferation
          regulation of proton transport
          negative regulation of epithelial cell migration
          positive regulation of B cell proliferation
          B cell costimulation
          positive regulation of interleukin-10 production
          positive regulation of interleukin-13 production
          T-helper 2 cell cytokine production
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          B cell activation
          positive regulation of tyrosine phosphorylation of Stat5 protein
          defense response to protozoan
          negative regulation of macrophage activation
          positive regulation of mast cell degranulation
          negative regulation of nitric oxide biosynthetic process
          T-helper 2 cell differentiation
          positive regulation of chemokine biosynthetic process
          positive regulation of MHC class II biosynthetic process
          positive regulation of T cell differentiation
          negative regulation of osteoclast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of isotype switching to IgE isotypes
          positive regulation of isotype switching to IgG isotypes
          regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of immune response
          negative regulation of T cell activation
          positive regulation of B cell activation
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of mononuclear cell migration
          dendritic cell differentiation
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of myoblast fusion
          negative regulation of complement-dependent cytotoxicity
          negative regulation of T-helper 17 cell differentiation
          negative regulation of endothelial cell apoptotic process
          positive regulation of eosinophil chemotaxis
          negative regulation of extrinsic apoptotic signaling pathway
          cytokine activity
          cytokine receptor binding
          interleukin-4 receptor binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-kp8dz:80/100.66.77.153:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline