Search
Search

Omp| Gene Symbol: Omp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011010.2 | NP_035140.1 | 1 | 437 | ||
| GenBank mRNA | AK133724.1 | - | 1 | 468 | ||
| BC046600.1 | - | 1 | 437 | 6312008 | ||
| BC058228.1 | - | 1 | 457 | |||
| BC132327.1 | - | 1 | 436 | 40130604 | ||
| BC138222.1 | - | 1 | 436 | 8861241 | ||