Search
Search

P2rx7| Gene Symbol: P2rx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038839.2 | NP_001033928.1 | 1 | 190 | ||
| NM_001038845.2 | NP_001033934.1 | 1 | 190 | |||
| NM_001038887.1 | NP_001033976.1 | 1 | 190 | |||
| NM_011027.3 | NP_035157.2 | 1 | 190 | |||
| GenBank mRNA | AJ489296.1 | - | 1 | 27 | ||
| AJ489297.1 | - | 1 | 27 | |||
| AK171132.1 | - | 1 | 162 | |||
| BC062808.1 | - | 1 | 92 | |||
| BC128303.1 | - | 1 | 34 | |||
| BC128304.1 | - | 1 | 34 | |||
| BC141120.1 | - | 1 | 57 | 9055732 | ||
| BY752747.1 | - | 1 | 132 | |||