Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKD1 SIRNA Assays ›
Gene Symbol:
Prkd1
Gene Name:
protein kinase D1
Gene Aliases:
PKD, PKD1, Pkcm, Prkcm
Chromosome Location:
Chr.12: 50341226 - 50649248 on Build GRCm38
UniGene ID:
Mm.133719
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS276345
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Prkd1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008858.3
NP_032884.2
6
1008
GenBank mRNA
Z34524.1
-
6
883
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
positive regulation of endothelial cell proliferation
immune system process
protein phosphorylation
apoptotic process
inflammatory response
Golgi organization
nervous system development
positive regulation of endothelial cell migration
regulation of keratinocyte proliferation
positive regulation of neuron projection development
phosphorylation
peptidyl-serine phosphorylation
cell differentiation
positive regulation of CREB transcription factor activity
positive regulation of peptidyl-serine phosphorylation
cellular response to oxidative stress
intracellular signal transduction
cellular response to vascular endothelial growth factor stimulus
positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of blood vessel endothelial cell migration
innate immune response
positive regulation of osteoblast differentiation
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
vascular endothelial growth factor receptor signaling pathway
Golgi vesicle transport
positive regulation of NF-kappaB transcription factor activity
regulation of release of sequestered calcium ion into cytosol
negative regulation of cell death
protein kinase D signaling
positive regulation of histone deacetylase activity
positive regulation of endothelial cell chemotaxis
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein kinase C activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
identical protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0