Search
Search

Psmd4| Gene Symbol: Psmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282017.1 | NP_001268946.1 | 8, 9 | 986 | ||
| NM_008951.2 | NP_032977.1 | 8, 9 | 977 | |||
| GenBank mRNA | AB029142.1 | - | 8, 9 | 908 | ||
| AB029143.1 | - | 8, 9 | 917 | |||
| AB029144.1 | - | 8, 9 | 915 | |||
| AF013099.1 | - | 8, 9 | 908 | |||
| AF175574.1 | - | 8, 9 | 917 | |||
| AK010969.1 | - | 8, 9 | 906 | |||
| AK145524.1 | - | 8, 9 | 925 | |||
| BC009005.1 | - | 8, 9 | 935 | 3581937 | ||
| BC016433.1 | - | 5, 6 | 1776 | |||