Search
Search

Psmd4| Gene Symbol: Psmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282017.1 | NP_001268946.1 | 2 | 176 | ||
| NM_008951.2 | NP_032977.1 | 2 | 176 | |||
| GenBank mRNA | AB029142.1 | - | 2 | 107 | ||
| AB029143.1 | - | 2 | 107 | |||
| AB029144.1 | - | 2 | 105 | |||
| AB029145.1 | - | 2 | 107 | |||
| AB029146.1 | - | 2 | 95 | |||
| AF013099.1 | - | 2 | 107 | |||
| AF175574.1 | - | 2 | 107 | |||
| AK010969.1 | - | 2 | 105 | |||
| AK145524.1 | - | 2 | 115 | |||
| BC009005.1 | - | 2 | 125 | 3581937 | ||
| BC016433.1 | - | 2 | 110 | |||
| BY099585.1 | - | 2 | 177 | |||