Search
Search

Rapgef6| Gene Symbol: Rapgef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252494.1 | NP_001239423.1 | 16 | 2263 | ||
| NM_001252496.1 | NP_001239425.1 | 16 | 2263 | |||
| NM_001252497.1 | NP_001239426.1 | 11 | 2521 | |||
| NM_175258.4 | NP_780467.2 | 16 | 2263 | |||
| GenBank mRNA | AK143331.1 | - | 4 | 650 | ||
| AK164343.1 | - | 11 | 2494 | |||
| AK220459.1 | - | 5 | 666 | |||
| BC141182.1 | - | 16 | 2162 | 9055794 | ||
| BC145503.1 | - | 16 | 2162 | |||