Search
Search

Ccl17| Gene Symbol: Ccl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011332.3 | NP_035462.2 | 3 | 294 | ||
| GenBank mRNA | AF125570.1 | - | 3 | 383 | ||
| AF125571.1 | - | 3 | 325 | |||
| AF125572.1 | - | 3 | 298 | |||
| AJ242587.1 | - | 3 | 273 | |||
| AK007663.1 | - | 3 | 295 | |||
| BC028505.1 | - | 3 | 278 | 3328278 | ||