Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SFRP1 SIRNA Assays ›
          Gene Symbol:
          Sfrp1
          Gene Name:
          secreted frizzled-related protein 1
          Gene Aliases:
          2210415K03Rik, AW011917, AW107218, AW742929, sFRP-1
          Chromosome Location:
          Chr.8: 23411382 - 23449632 on Build GRCm38
          UniGene ID:
          Mm.281691
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS277028
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sfrp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013834.3 NP_038862.2 3 1012
          GenBank mRNA AK081052.1 - 3 603
          AK088868.1 - 3 1014
          BC024495.1 - 3 982 4982195
          BC094662.1 - 3 996 30363219

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          somitogenesis
          neural tube closure
          hematopoietic progenitor cell differentiation
          proteolysis
          multicellular organism development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          male gonad development
          female gonad development
          dorsal/ventral axis specification
          anterior/posterior pattern specification
          regulation of cell cycle process
          negative regulation of gene expression
          negative regulation of epithelial to mesenchymal transition
          neural crest cell fate commitment
          response to organic cyclic compound
          Wnt signaling pathway
          neural tube development
          actin cytoskeleton organization
          hemopoiesis
          cell differentiation
          positive regulation of Wnt signaling pathway
          negative regulation of Wnt signaling pathway
          regulation of ossification
          negative regulation of ossification
          positive regulation of cell growth
          negative regulation of cell growth
          negative regulation of cell migration
          negative regulation of BMP signaling pathway
          negative regulation of osteoblast proliferation
          substrate adhesion-dependent cell spreading
          somatic stem cell population maintenance
          response to drug
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of JUN kinase activity
          cellular response to fibroblast growth factor stimulus
          negative regulation of B cell differentiation
          positive regulation of fat cell differentiation
          negative regulation of osteoblast differentiation
          negative regulation of osteoclast differentiation
          positive regulation of smoothened signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          development of primary male sexual characteristics
          negative regulation of insulin secretion
          negative regulation of bone remodeling
          negative regulation of fibroblast proliferation
          digestive tract morphogenesis
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          negative regulation of peptidyl-tyrosine phosphorylation
          canonical Wnt signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          hematopoietic stem cell differentiation
          bone trabecula formation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          regulation of branching involved in prostate gland morphogenesis
          negative regulation of androgen receptor signaling pathway
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          cellular response to estrogen stimulus
          cellular response to X-ray
          cellular response to heparin
          negative regulation of canonical Wnt signaling pathway
          regulation of establishment of planar polarity
          planar cell polarity pathway involved in neural tube closure
          Wnt signaling pathway involved in somitogenesis
          convergent extension involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of planar cell polarity pathway involved in axis elongation
          positive regulation of non-canonical Wnt signaling pathway
          negative regulation of Wnt signaling pathway involved in dorsal/ventral axis specification
          negative regulation of canonical Wnt signaling pathway involved in controlling type B pancreatic cell proliferation
          negative regulation of fibroblast apoptotic process
          positive regulation of fibroblast apoptotic process
          positive regulation of extrinsic apoptotic signaling pathway
          cysteine-type endopeptidase activity
          frizzled binding
          protein binding
          drug binding
          heparin binding
          Wnt-protein binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0