Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SORL1 SIRNA Assays ›
          Gene Symbol:
          Sorl1
          Gene Name:
          sortilin-related receptor, LDLR class A repeats-containing
          Gene Aliases:
          2900010L19Rik, AI596264, AW261561, LR11, SorLA, gp250, mSorLA
          Chromosome Location:
          Chr.9: 41968489 - 42124289 on Build GRCm38
          UniGene ID:
          Mm.121920
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS277176
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sorl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011436.3 NP_035566.2 33, 34 4833
          GenBank mRNA AB015790.1 - 33, 34 4657
          AF031816.1 - 4111
          AK137635.1 - 21, 22 2948
          AK147303.1 - 33, 34 4834
          Y12004.1 - 10, 11 1303

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to Golgi
          protein targeting
          protein targeting to lysosome
          lipid metabolic process
          transport
          lipid transport
          post-Golgi vesicle-mediated transport
          endocytosis
          multicellular organism development
          steroid metabolic process
          cholesterol metabolic process
          regulation of smooth muscle cell migration
          cell migration
          negative regulation of protein binding
          negative regulation of protein oligomerization
          negative regulation of MAP kinase activity
          protein retention in Golgi apparatus
          positive regulation of protein catabolic process
          negative regulation of neurogenesis
          protein maturation
          positive regulation of protein exit from endoplasmic reticulum
          negative regulation of neuron death
          negative regulation of beta-amyloid formation
          positive regulation of choline O-acetyltransferase activity
          negative regulation of tau-protein kinase activity
          positive regulation of ER to Golgi vesicle-mediated transport
          positive regulation of early endosome to recycling endosome transport
          negative regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
          positive regulation of protein localization to early endosome
          negative regulation of neurofibrillary tangle assembly
          positive regulation of endocytic recycling
          beta-amyloid binding
          protein binding
          low-density lipoprotein particle binding
          ADP-ribosylation factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0