Search
Search

Mlx| Gene Symbol: Mlx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159384.1 | NP_001152856.1 | 3 | 207 | ||
| NM_001159385.1 | NP_001152857.1 | 4 | 297 | |||
| NM_011550.3 | NP_035680.3 | 4 | 459 | |||
| GenBank mRNA | AF213670.1 | - | 3 | 171 | ||
| AF213671.1 | - | 4 | 261 | |||
| AF213672.1 | - | 4 | 423 | |||
| AK011478.1 | - | 3 | 187 | |||
| AK142605.1 | - | 4 | 263 | |||
| BC031380.1 | - | 4 | 439 | 5252695 | ||
| BC038004.1 | - | 4 | 439 | |||
| BC080768.1 | - | 3 | 1264 | |||
| U43548.1 | - | 4 | 419 | |||
| U43607.1 | - | 4 | 430 | |||