Search
Search

Tgfbr2| Gene Symbol: Tgfbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009371.3 | NP_033397.3 | 5 | 940 | ||
| NM_029575.3 | NP_083851.3 | 4 | 865 | |||
| GenBank mRNA | AF406755.1 | - | 4 | 539 | ||
| AK043619.1 | - | 5 | 1024 | |||
| AK046561.1 | - | 5 | 866 | |||
| AK053793.1 | - | 5 | 941 | |||
| AK148167.1 | - | 4 | 865 | |||
| AK159327.1 | - | 5 | 920 | |||
| BC052629.1 | - | 842 | ||||
| S69114.1 | - | 4 | 576 | |||