Search
Search

Top3a| Gene Symbol: Top3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009410.2 | NP_033436.1 | 8 | 1113 | ||
| GenBank mRNA | AB006074.1 | - | 8 | 1114 | ||
| AK029987.1 | - | 5 | 506 | |||
| AK037640.1 | - | 8 | 1014 | |||
| AK164443.1 | - | 8 | 1014 | |||
| AK170291.1 | - | 9 | 920 | |||
| AK171286.1 | - | 9 | 920 | |||
| BC099689.1 | - | 8 | 928 | 30731611 | ||