Search
Search

Dctn6| Gene Symbol: Dctn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293757.1 | NP_001280686.1 | 4 | 531 | ||
| NM_001293758.1 | NP_001280687.1 | 3 | 364 | |||
| NM_001293759.1 | NP_001280688.1 | 4 | 601 | |||
| NM_011722.4 | NP_035852.1 | 3 | 418 | |||
| GenBank mRNA | AF124788.1 | - | 3 | 149 | ||
| AF190796.1 | - | 3 | 159 | |||
| AK003618.1 | - | 3 | 158 | |||
| AK088312.1 | - | 3 | 165 | |||
| BC029249.1 | - | 3 | 159 | 4486579 | ||
| BY079663.1 | - | 3 | 218 | |||
| CV560309.1 | - | 3 | 315 | |||
| DV042569.1 | - | 3 | 373 | |||
| DV062065.1 | - | 4 | 323 | |||