Search
Search

Eci2| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110331.1 | NP_001103801.1 | 2 | 199 | ||
| NM_011868.3 | NP_035998.2 | 2 | 383 | |||
| GenBank mRNA | AF153613.1 | - | 2 | 355 | ||
| AK170778.1 | - | 2 | 168 | |||
| AK182614.1 | - | 2 | 109 | |||
| BC001983.1 | - | 2 | 93 | 3591615 | ||
| BY037058.1 | - | 2 | 199 | |||
| BY238570.1 | - | 2 | 125 | |||
| BY251531.1 | - | 2 | 366 | |||
| CB318094.1 | - | 2 | 180 | |||
| CX242291.1 | - | 2 | 362 | |||