Search
Search

Zdhhc19| Gene Symbol: Zdhhc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199309.2 | NP_955013.1 | 4, 5 | 540 | ||
| GenBank mRNA | AK132780.1 | - | 4, 5 | 557 | ||
| AK133398.1 | - | 4, 5 | 541 | |||
| BC049761.1 | - | 4, 5 | 540 | 6773873 | ||
| BC141059.1 | - | 3, 4 | 449 | 9055671 | ||