Search
Search

Wfdc16| Gene Symbol: Wfdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012723.2 | NP_001012741.2 | 4 | 396 | ||
| GenBank mRNA | AY542488.1 | - | 4 | 396 | ||
| AY542489.1 | - | 4 | 396 | |||
| BC043944.1 | - | 4 | 390 | |||
| BC145970.1 | - | 3 | 343 | 40131273 | ||
| BC145972.1 | - | 3 | 343 | 40131275 | ||