Search
Search

Lrrc58| Gene Symbol: Lrrc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177093.3 | NP_796067.2 | 1, 2 | 580 | ||
| GenBank mRNA | AK049274.1 | - | 1, 2 | 581 | ||
| AK090204.1 | - | 1, 2 | 590 | |||
| AK147818.1 | - | 1, 2 | 556 | |||
| BC141007.1 | - | 1, 2 | 540 | 9055619 | ||