Search
Search

Lair1| Gene Symbol: Lair1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113474.1 | NP_001106945.1 | 7 | 727 | ||
| NM_178611.6 | NP_848726.1 | 6 | 397 | |||
| GenBank mRNA | AF479685.1 | - | 7 | 662 | ||
| AF491829.1 | - | 7 | 760 | |||
| AF491830.1 | - | 6 | 309 | |||
| AF491831.1 | - | 7 | 639 | |||
| AK017222.1 | - | 6 | 401 | |||
| AK033472.1 | - | 6 | 374 | |||
| AK038574.1 | - | 6 | 360 | |||
| AK043664.1 | - | 6 | 454 | |||
| AK044465.1 | - | 7 | 698 | |||
| AK050281.1 | - | 7 | 683 | |||
| AY392763.1 | - | 7 | 664 | |||
| AY392764.1 | - | 6 | 334 | |||
| AY392765.1 | - | 5 | 217 | |||
| AY392766.1 | - | 6 | 547 | |||
| BC025901.1 | - | 7 | 691 | 5253846 | ||
| BY204263.1 | - | 5 | 341 | |||