Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM3 SIRNA Assays ›
Gene Symbol:
Tpm3
Gene Name:
tropomyosin 3, gamma
Gene Aliases:
TM30nm, TMnm, Tm5NM, Tpm-5, Tpm5, Trop-5, gamma-TM, hTM30nm, hTMnm
Chromosome Location:
Chr.3: 90072651 - 90100902 on Build GRCm38
UniGene ID:
Mm.240839
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS286250
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253738.1
NP_001240667.1
3
535
NM_001253740.1
NP_001240669.1
3
535
NM_001271764.1
NP_001258693.1
3
535
NM_001293748.1
NP_001280677.2
4
518
NM_001293749.1
NP_001280678.1
3
535
NM_022314.3
NP_071709.3
4
518
GenBank mRNA
AF317223.1
-
4
432
AK050665.1
-
3
381
AK088111.1
-
3
395
AK088902.1
-
3
393
AK145238.1
-
3
392
AK145243.1
-
3
392
AK145804.1
-
3
392
AK146349.1
-
3
365
AK146557.1
-
3
517
AK151441.1
-
3
417
AK151500.1
-
3
392
AK151571.1
-
3
391
AK151578.1
-
3
392
AK151595.1
-
3
364
AK151602.1
-
3
392
AK151991.1
-
3
391
AK152260.1
-
3
391
AK152659.1
-
3
364
AK153031.1
-
3
364
AK160011.1
-
3
417
AK164086.1
-
3
392
AK165428.1
-
3
391
AK166836.1
-
3
379
AK167268.1
-
3
369
AK167534.1
-
3
397
AK167581.1
-
3
392
AK168843.1
-
3
360
AK169102.1
-
3
363
AK169245.1
-
3
368
AK169821.1
-
3
409
BC029186.1
-
4
475
4502880
BC055407.1
-
3661
BC092045.1
-
3
368
3591372
CX237327.1
-
3
357
U04541.1
-
4
488
X53753.1
-
3
403
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Process(es)
actin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0