Search
Search

Nrarp| Gene Symbol: Nrarp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025980.2 | NP_080256.2 | 1 | 554 | ||
| GenBank mRNA | AK012426.1 | - | 1 | 289 | ||
| AY046077.1 | - | 1 | 554 | |||
| BC020293.1 | - | 1 | 551 | |||
| BC048088.1 | - | 1 | 531 | |||
| BC069891.1 | - | 1 | 307 | 5391567 | ||