Search
Search

Exosc8| Gene Symbol: Exosc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163570.1 | NP_001157042.1 | 6 | 663 | ||
| NM_027148.3 | NP_081424.3 | 6 | 651 | |||
| GenBank mRNA | AA003291.1 | - | 6 | 338 | ||
| AJ505005.1 | - | 6 | 311 | |||
| AK009584.1 | - | 6 | 323 | |||
| AK047257.1 | - | 5 | 1136 | |||
| AK169719.1 | - | 4 | 254 | |||
| AK220227.1 | - | 6 | 651 | |||
| BC004769.1 | - | 5 | 806 | |||
| BC059089.1 | - | 6 | 680 | 6509820 | ||
| BC067250.1 | - | 5 | 294 | |||