Search
Search

Metrn| Gene Symbol: Metrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133719.2 | NP_598480.1 | 2 | 372 | ||
| GenBank mRNA | AK007688.1 | - | 2 | 312 | ||
| AK087402.1 | - | 2 | 373 | |||
| BC008695.1 | - | 2 | 250 | |||
| BC037181.1 | - | 2 | 299 | |||
| BC132177.1 | - | 3 | 374 | 40130454 | ||
| DQ133462.1 | - | 2 | 295 | |||