Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SASH1 SIRNA Assays ›
Gene Symbol:
Sash1
Gene Name:
SAM and SH3 domain containing 1
Gene Aliases:
1100001C18Rik, 2500002E12Rik, A330076K04Rik, mKIAA0790
Chromosome Location:
Chr.10: 8722217 - 8956137 on Build GRCm38
UniGene ID:
Mm.338275
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS291087
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sash1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_175155.4
NP_780364.3
6
689
GenBank mRNA
AJ507736.1
-
6
504
AK129216.1
-
1
23
AK149872.1
-
6
641
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_015278, AJ507735, AB018333.
This siRNA also targets the following transcripts in Rat: XM_003748645.
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
positive regulation of endothelial cell migration
positive regulation of lipopolysaccharide-mediated signaling pathway
positive regulation of JUN kinase activity
positive regulation of angiogenesis
regulation of protein K63-linked ubiquitination
positive regulation of p38MAPK cascade
positive regulation of NIK/NF-kappaB signaling
regulation of protein autoubiquitination
protein C-terminus binding
protein kinase binding
mitogen-activated protein kinase kinase kinase binding
protein complex scaffold
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline