Search
Search

Smad1| Gene Symbol: Smad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013130.2 | NP_037262.2 | 7 | 1724 | ||
| XM_006255394.1 | XP_006255456.1 | 7 | 1870 | |||
| XM_006255395.2 | XP_006255457.1 | 7 | 2529 | |||
| XM_006255396.2 | XP_006255458.1 | 7 | 1760 | |||
| XM_006255397.1 | XP_006255459.1 | 7 | 1849 | |||
| GenBank mRNA | AF067727.1 | - | 6 | 1405 | ||
| BC061757.1 | - | 7 | 1724 | 5599977 | ||
| U66478.1 | - | 7 | 1705 | |||