Search
Search

Sema3f| Gene Symbol: Sema3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108185.1 | NP_001101655.1 | 3 | 346 | ||
| XM_006243778.2 | XP_006243840.1 | 3 | 901 | |||
| XM_008766532.1 | XP_008764754.1 | 3 | 229 | |||
| XM_008766533.1 | XP_008764755.1 | 1 | 1283 | |||