Search
Search

Gramd1a| Gene Symbol: Gramd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014160.1 | NP_001014182.1 | 13 | 1536 | ||
| XM_006228832.2 | XP_006228894.1 | 15 | 1951 | |||
| XM_006228833.2 | XP_006228895.1 | 15 | 1949 | |||
| XM_006228834.2 | XP_006228896.1 | 14 | 1717 | |||
| XM_006228835.2 | XP_006228897.1 | 14 | 1715 | |||
| GenBank mRNA | AY171575.1 | - | 13 | 1536 | ||
| BC105896.1 | - | 14 | 1637 | 7380410 | ||