Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TP73 SIRNA Assays ›
Gene Symbol:
Tp73
Gene Name:
tumor protein p73
Gene Aliases:
P73, Trp73
Chromosome Location:
Chr.5: 171355876 - 171415354 on Build Rnor_6.0
UniGene ID:
Rn.103860
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS325328
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tp73
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001108696.1
NP_001102166.1
9
1163
Back To Top
More Information
Panther Classification:
Molecular Function -
P53-like transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
activation of MAPK activity
kidney development
release of cytochrome c from mitochondria
transcription from RNA polymerase II promoter
inflammatory response
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
cell cycle arrest
regulation of mitotic cell cycle
post-embryonic development
response to X-ray
response to organonitrogen compound
response to gamma radiation
regulation of gene expression
hippocampus development
forebrain development
mitotic G1 DNA damage checkpoint
cerebrospinal fluid secretion
cellular response to UV
response to drug
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of apoptotic process
negative regulation of JUN kinase activity
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
negative regulation of neuron differentiation
positive regulation of cell size
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
digestive tract morphogenesis
neuron development
positive regulation of oligodendrocyte differentiation
protein tetramerization
negative regulation of cardiac muscle cell proliferation
positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor binding
protein kinase binding
identical protein binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0