Search
Search

Dixdc1| Gene Symbol: Dixdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037654.1 | NP_001032743.1 | 6 | 767 | ||
| XM_006243042.2 | XP_006243104.1 | 6 | 1224 | |||
| XM_006243045.2 | XP_006243107.1 | 5 | 579 | |||
| XM_008766295.1 | XP_008764517.1 | 6 | 1260 | |||
| XM_008766296.1 | XP_008764518.1 | 2 | 591 | |||
| GenBank mRNA | AY770507.1 | - | 6 | 767 | ||