Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC103689963 SIRNA Assays ›
          Gene Symbol:
          LOC103689963 Hdac6
          Gene Name:
          histone deacetylase 6
          histone deacetylase 6
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.X: 15295388 - 15316673 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 15396201 - 15417486 on Build Rnor_6.0
          UniGene ID:
          -
          Rn.13453
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID RSS331540
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC103689963
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006256755.2 XP_006256817.1 4 560
          XM_006256756.2 XP_006256818.1 4 354
          XM_228753.8 XP_228753.4 4 485
          Gene Symbol: Hdac6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003752010.3 XP_003752058.1 485
          XM_006256758.2 XP_006256820.1 560
          XM_006256759.2 XP_006256821.1 354

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          regulation of protein phosphorylation
          response to amphetamine
          protein deacetylation
          ubiquitin-dependent protein catabolic process
          misfolded or incompletely synthesized protein catabolic process
          intracellular protein transport
          negative regulation of microtubule depolymerization
          metabolic process
          positive regulation of cell proliferation
          response to toxic substance
          positive regulation of signal transduction
          response to organic substance
          regulation of receptor activity
          positive regulation of epithelial cell migration
          regulation of mitochondrion organization
          positive regulation of receptor biosynthetic process
          negative regulation of neuron projection development
          macroautophagy
          histone deacetylation
          axonal transport of mitochondrion
          neuron differentiation
          lysosome localization
          cellular response to oxidative stress
          peptidyl-lysine deacetylation
          response to immobilization stress
          cellular response to topologically incorrect protein
          regulation of gene expression, epigenetic
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          protein complex disassembly
          negative regulation of protein complex disassembly
          regulation of fat cell differentiation
          negative regulation of proteolysis
          negative regulation of axon extension involved in axon guidance
          positive regulation of dendrite morphogenesis
          response to corticosterone
          response to misfolded protein
          positive regulation of synaptic transmission, glutamatergic
          regulation of establishment of protein localization
          cellular response to hydrogen peroxide
          regulation of microtubule cytoskeleton organization
          aggresome assembly
          polyubiquitinated misfolded protein transport
          Hsp90 deacetylation
          response to growth factor
          cellular response to misfolded protein
          cellular response to parathyroid hormone stimulus
          response to dexamethasone
          positive regulation of chaperone-mediated protein complex assembly
          tubulin deacetylation
          positive regulation of tubulin deacetylation
          positive regulation of hydrogen peroxide-mediated programmed cell death
          negative regulation of protein acetylation
          positive regulation of oxidative stress-induced neuron death
          core promoter binding
          histone deacetylase activity
          protein binding
          beta-catenin binding
          microtubule binding
          zinc ion binding
          hydrolase activity
          polyubiquitin binding
          ubiquitin protein ligase binding
          histone deacetylase binding
          tubulin deacetylase activity
          alpha-tubulin binding
          ubiquitin binding
          tau protein binding
          beta-tubulin binding
          misfolded protein binding
          Hsp90 protein binding
          dynein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0