Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOD1 SIRNA Assays ›
          Gene Symbol:
          Sod1
          Gene Name:
          superoxide dismutase 1, soluble
          Gene Aliases:
          CuZnSOD
          Chromosome Location:
          Chr.11: 30363282 - 30368858 on Build Rnor_6.0
          UniGene ID:
          Rn.6059
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS332482
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sod1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017050.1 NP_058746.1 5 485
          GenBank mRNA BC082800.1 - 5 465 7304983
          FQ209495.1 - 5 486
          FQ209941.1 - 5 481
          FQ220715.1 - 5 485
          M21060.1 - 5 388
          M25157.1 - 5 444
          X05634.1 - 5 384
          Y00404.1 - 5 485

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_000454, X02317, AV756797, AY049787, CR450355, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011434, AK077284, BC002066, X06683, AK020624, and others.

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          response to reactive oxygen species
          response to superoxide
          ovarian follicle development
          positive regulation of cytokine production
          retina homeostasis
          response to amphetamine
          myeloid cell homeostasis
          double-strand break repair
          apoptotic DNA fragmentation
          glutathione metabolic process
          superoxide metabolic process
          cellular iron ion homeostasis
          response to oxidative stress
          spermatogenesis
          embryo implantation
          aging
          cell aging
          sensory perception of sound
          locomotory behavior
          anterograde axonal transport
          retrograde axonal transport
          regulation of blood pressure
          response to heat
          response to organic substance
          transmission of nerve impulse
          removal of superoxide radicals
          response to nutrient levels
          peripheral nervous system myelin maintenance
          positive regulation of superoxide anion generation
          response to carbon monoxide
          cellular response to oxidative stress
          cellular response to potassium ion
          regulation of multicellular organism growth
          response to drug
          response to hydrogen peroxide
          superoxide anion generation
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          negative regulation of neuron apoptotic process
          response to ethanol
          negative regulation of cholesterol biosynthetic process
          regulation of protein kinase activity
          response to antibiotic
          response to copper ion
          muscle cell cellular homeostasis
          response to axon injury
          hydrogen peroxide biosynthetic process
          regulation of mitochondrial membrane potential
          oxidation-reduction process
          heart contraction
          neurofilament cytoskeleton organization
          relaxation of vascular smooth muscle
          auditory receptor cell stereocilium organization
          cellular response to cadmium ion
          cellular response to ATP
          reactive oxygen species metabolic process
          response to antipsychotic drug
          positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          superoxide dismutase activity
          copper ion binding
          zinc ion binding
          enzyme binding
          protein phosphatase 2B binding
          identical protein binding
          Rac GTPase binding
          chaperone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0