Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VDR SIRNA Assays ›
Gene Symbol:
Vdr
Gene Name:
vitamin D (1,25- dihydroxyvitamin D3) receptor
Gene Aliases:
Nr1i1
Chromosome Location:
Chr.7: 139344452 - 139394138 on Build Rnor_6.0
UniGene ID:
Rn.10911
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS332506
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Vdr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017058.1
NP_058754.1
5
401
GenBank mRNA
J04147.1
-
5
401
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
skeletal system development
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
calcium ion transport
cellular calcium ion homeostasis
multicellular organism development
heart development
aging
lactation
negative regulation of cell proliferation
organ morphogenesis
positive regulation of gene expression
negative regulation of keratinocyte proliferation
positive regulation of vitamin D 24-hydroxylase activity
intracellular receptor signaling pathway
response to estradiol
bile acid signaling pathway
steroid hormone mediated signaling pathway
positive regulation of keratinocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
decidualization
intestinal absorption
response to calcium ion
regulation of calcium ion transport
positive regulation of apoptotic process involved in mammary gland involution
regulation of calcidiol 1-monooxygenase activity
mammary gland branching involved in pregnancy
vitamin D receptor signaling pathway
cellular response to vitamin D
apoptotic signaling pathway
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
vitamin D binding
protein binding
zinc ion binding
calcitriol receptor activity
lithocholic acid receptor activity
sequence-specific DNA binding
retinoid X receptor binding
vitamin D response element binding
calcitriol binding
lithocholic acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline