Search
Search

Akr7a3| Gene Symbol: Akr7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013215.1 | NP_037347.1 | 5 | 675 | ||
| GenBank mRNA | AF045464.1 | - | 5 | 675 | ||
| BC078872.1 | - | 5 | 669 | 7127297 | ||
| BC089811.1 | - | 5 | 638 | 7126881 | ||
| X74673.1 | - | 5 | 672 | |||