Search
Search

LOC103690048
Klk1c8| Gene Symbol: LOC103690048 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759444.1 | XP_008757666.1 | 2 | 103 | ||
| Gene Symbol: Klk1c8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013067.2 | NP_001013085.1 | 2 | 64 | ||