Search
Search

Drap1| Gene Symbol: Drap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077668.2 | NP_001071136.1 | 7 | 804 | ||
| XM_006230741.1 | XP_006230803.1 | 7 | 868 | |||
| GenBank mRNA | BC127524.1 | - | 7 | 805 | 7114598 | |
| FQ231535.1 | - | 7 | 691 | |||