Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other G6PD SIRNA Assays ›
Gene Symbol:
G6pd
Gene Name:
glucose-6-phosphate dehydrogenase
Gene Aliases:
G6pdx
Chromosome Location:
Chr.X: 156274800 - 156293935 on Build Rnor_6.0
UniGene ID:
Rn.11040
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS351259
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
458.00
Your Price
Online offer:
458.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: G6pd
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017006.2
NP_058702.1
11
1440
GenBank mRNA
BC081820.1
-
11
1353
7108556
X07467.1
-
11
1366
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019468, NM_008062, AK165207, Z11911, AK088135, and others.
Panther Classification:
Molecular Function -
dehydrogenase
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure
vasodilation by angiotensin involved in regulation of systemic arterial blood pressure
glucose metabolic process
pentose-phosphate shunt
cholesterol biosynthetic process
NADP metabolic process
NADPH regeneration
NADP biosynthetic process
glutathione metabolic process
response to oxidative stress
pentose-phosphate shunt, oxidative branch
negative regulation of protein glutathionylation
response to organic cyclic compound
pentose biosynthetic process
substantia nigra development
response to food
interleukin-10 production
interleukin-12 production
cellular response to oxidative stress
regulation of multicellular organism growth
erythrocyte maturation
regulation of neuron apoptotic process
response to ethanol
ribose phosphate biosynthetic process
erythrocyte development
glucose 6-phosphate metabolic process
oxidation-reduction process
glucose-6-phosphate dehydrogenase activity
glucose binding
carbohydrate binding
identical protein binding
protein homodimerization activity
NADP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0