Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GSTP1 SIRNA Assays ›
          Gene Symbol:
          Gstp1
          Gene Name:
          glutathione S-transferase pi 1
          Gene Aliases:
          GST-P, Gst3, Gstp, Gstp2
          Chromosome Location:
          Chr.1: 219291679 - 219294147 on Build Rnor_6.0
          UniGene ID:
          Rn.87063
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS351296
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gstp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012577.2 NP_036709.1 7 688
          GenBank mRNA BC058439.1 - 7 692 6889675
          BC058440.1 - 7 692 6917961
          FJ179407.1 - 6 618
          FQ217279.1 - 7 688

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          negative regulation of protein kinase activity
          glutathione metabolic process
          xenobiotic metabolic process
          response to toxic substance
          negative regulation of biosynthetic process
          oligodendrocyte development
          organ regeneration
          response to nutrient levels
          response to estradiol
          negative regulation of interleukin-1 beta production
          negative regulation of tumor necrosis factor production
          cellular response to insulin stimulus
          regulation of stress-activated MAPK cascade
          negative regulation of stress-activated MAPK cascade
          positive regulation of superoxide anion generation
          response to L-ascorbic acid
          common myeloid progenitor cell proliferation
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to amino acid
          negative regulation of MAP kinase activity
          negative regulation of JUN kinase activity
          response to ethanol
          response to mercury ion
          negative regulation of fibroblast proliferation
          negative regulation of nitric-oxide synthase biosynthetic process
          regulation of ERK1 and ERK2 cascade
          negative regulation of ERK1 and ERK2 cascade
          negative regulation of leukocyte proliferation
          cellular response to lipopolysaccharide
          cellular response to epidermal growth factor stimulus
          cellular response to glucocorticoid stimulus
          cellular response to cell-matrix adhesion
          negative regulation of monocyte chemotactic protein-1 production
          negative regulation of peroxidase activity
          negative regulation of extrinsic apoptotic signaling pathway
          glutathione transferase activity
          drug binding
          JUN kinase binding
          kinase regulator activity
          S-nitrosoglutathione binding
          dinitrosyl-iron complex binding
          glutathione binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0