Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AHR SIRNA Assays ›
Gene Symbol:
Ahr
Gene Name:
aryl hydrocarbon receptor
Gene Aliases:
-
Chromosome Location:
Chr.6: 54966533 - 55001436 on Build Rnor_6.0
UniGene ID:
Rn.91370
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS352343
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ahr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013149.2
NP_037281.2
10
2212
GenBank mRNA
U09000.1
-
10
2212
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
ovarian follicle development
blood vessel development
patterning of blood vessels
B cell homeostasis
liver development
blood vessel remodeling
lymphocyte homeostasis
immune system process
negative regulation of systemic arterial blood pressure
cardiac left ventricle morphogenesis
circumferential growth involved in left ventricle morphogenesis
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
apoptotic process
blood circulation
response to xenobiotic stimulus
response to toxic substance
regulation of gene expression
response to organic cyclic compound
intracellular receptor signaling pathway
prostate gland development
regulation of B cell proliferation
response to estradiol
circadian regulation of gene expression
embryonic hemopoiesis
post-embryonic hemopoiesis
camera-type eye development
T cell homeostasis
positive regulation of cell size
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
negative regulation of vasoconstriction
positive regulation of growth
positive regulation of transcription from RNA polymerase II promoter
blood vessel morphogenesis
spleen development
reproductive structure development
gland development
smooth muscle tissue development
regulation of blood vessel size
negative regulation of necrotic cell death
venous blood vessel development
kidney morphogenesis
common bile duct development
cellular response to cAMP
cellular response to glucocorticoid stimulus
glomerulus morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
signal transducer activity
aryl hydrocarbon receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
transcription factor binding
enhancer binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
protein dimerization activity
Hsp90 protein binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0