Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS3A SIRNA Assays ›
          Gene Symbol:
          Rps3a Cyld
          Gene Name:
          ribosomal protein S3a
          cylindromatosis (turban tumor syndrome)
          Gene Aliases:
          -
          LRRGT00003, Rp1, Rp1h
          Chromosome Location:
          Chr.2: 185440477 - 185444846 on Build Rnor_6.0
          Chromosome Location:
          Chr.19: 19260506 - 19323817 on Build Rnor_6.0
          UniGene ID:
          Rn.94935
          Rn.128760
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID RSS354408
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps3a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017153.1 NP_058849.1 4 567
          GenBank mRNA BC058483.1 - 4 572 6887884
          FQ209727.1 - 4 567
          FQ211699.1 - 4 568
          FQ211708.1 - 4 567
          FQ216850.1 - 4 567
          FQ217809.1 - 4 567
          FQ219020.1 - 4 567
          FQ219522.1 - 4 568
          FQ219867.1 - 4 568
          FQ220002.1 - 4 567
          FQ220027.1 - 4 567
          FQ220037.1 - 4 568
          FQ220045.1 - 4 567
          FQ220135.1 - 4 568
          FQ220260.1 - 4 567
          FQ220261.1 - 4 568
          FQ220317.1 - 4 567
          FQ220504.1 - 4 567
          FQ220529.1 - 4 567
          FQ220577.1 - 4 568
          FQ220731.1 - 4 567
          FQ221431.1 - 4 568
          FQ221460.1 - 4 567
          FQ221744.1 - 4 567
          FQ221931.1 - 4 567
          FQ222056.1 - 4 567
          FQ222736.1 - 4 568
          FQ222773.1 - 4 568
          FQ222846.1 - 4 597
          FQ222869.1 - 4 567
          FQ222876.1 - 4 567
          FQ223535.1 - 4 574
          FQ223540.1 - 4 567
          FQ225080.1 - 4 568
          FQ225145.1 - 4 567
          FQ225713.1 - 4 566
          FQ227696.1 - 4 567
          FQ227805.1 - 4 568
          FQ227878.1 - 4 568
          FQ228312.1 - 4 568
          FQ228317.1 - 4 567
          FQ228376.1 - 4 568
          FQ228558.1 - 4 567
          FQ229234.1 - 4 567
          FQ229381.1 - 4 568
          FQ229520.1 - 4 567
          FQ229870.1 - 4 567
          FQ229960.1 - 4 567
          FQ231162.1 - 4 568
          FQ231673.1 - 4 567
          FQ232329.1 - 4 569
          FQ232836.1 - 4 567
          FQ232928.1 - 4 568
          FQ233768.1 - 4 571
          FQ234311.1 - 4 567
          FQ234537.1 - 4 567
          M84716.1 - 4 561
          X75161.1 - 4 567
          Gene Symbol: Cyld
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY383658.1 - 1 525

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoplasmic translation
          translation
          biological_process
          cell differentiation
          negative regulation of apoptotic process
          molecular_function
          structural constituent of ribosome
          translation initiation factor binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0