Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RT1-DB1 SIRNA Assays ›
          Gene Symbol:
          RT1-Db1
          Gene Name:
          RT1 class II, locus Db1
          Gene Aliases:
          Db1, RT1-Db, RT1-Db1n
          Chromosome Location:
          Chr.20: 4087621 - 4097190 on Build Rnor_6.0
          UniGene ID:
          Rn.33311
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS354971
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RT1-Db1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008884.2 NP_001008884.1 2, 3 418
          GenBank mRNA AF084933.1 - 2, 3 358
          AJ003231.1 - 2, 3 303
          AJ003234.1 - 2, 3 303
          AY626207.1 - 2, 3 354
          BC087691.1 - 3, 4 338 7310949
          CO563535.1 - 2, 3 418
          FQ225193.1 - 2, 3 406
          FQ226724.1 - 2, 3 406
          FQ226759.1 - 2, 3 412
          FQ226850.1 - 2, 3 406
          FQ227087.1 - 2, 3 401
          FQ228005.1 - 2, 3 406
          FQ228124.1 - 2, 3 383
          FQ228129.1 - 2, 3 406
          FQ228185.1 - 2, 3 424
          FQ230360.1 - 2, 3 406
          FQ231387.1 - 2, 3 406
          FQ231709.1 - 2, 3 406
          FQ232046.1 - 2, 3 406
          FQ232100.1 - 2, 3 406
          FQ232453.1 - 2, 3 406
          FQ233049.1 - 2, 3 402
          FQ233335.1 - 2, 3 406
          FQ233396.1 - 2, 3 406
          FQ233493.1 - 2, 3 406
          FQ233801.1 - 2, 3 406
          FQ234464.1 - 2, 3 405
          FQ234907.1 - 2, 3 406
          KC222950.1 - 2, 3 354

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immunoglobulin production involved in immunoglobulin mediated immune response
          inflammatory response to antigenic stimulus
          humoral immune response mediated by circulating immunoglobulin
          peptide antigen assembly with MHC class II protein complex
          antigen processing and presentation of peptide or polysaccharide antigen via MHC class II
          polysaccharide assembly with MHC class II protein complex
          negative regulation of inflammatory response to antigenic stimulus
          immune response
          biological_process
          detection of bacterium
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          regulation of interleukin-4 production
          negative regulation of interferon-gamma production
          response to interferon-gamma
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          T-helper 1 type immune response
          negative regulation of T cell proliferation
          protein tetramerization
          regulation of interleukin-10 secretion
          molecular_function
          MHC class II protein complex binding
          peptide antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0