Search
Search

Sbf2| Gene Symbol: Sbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134970.1 | NP_001128442.1 | 15, 16 | 1816 | ||
| XM_008759797.1 | XP_008758019.1 | 16, 17 | 2032 | |||
| XM_008759798.1 | XP_008758020.1 | 16, 17 | 2032 | |||
| XM_008759799.1 | XP_008758021.1 | 16, 17 | 2032 | |||
| XM_008759800.1 | XP_008758022.1 | 16, 17 | 2032 | |||
| XM_008759801.1 | XP_008758023.1 | 16, 17 | 2032 | |||
| XM_008759802.1 | XP_008758024.1 | 15, 16 | 1894 | |||
| XM_008759803.1 | XP_008758025.1 | 15, 16 | 1812 | |||
| XM_008759804.1 | XP_008758026.1 | 16, 17 | 2032 | |||
| GenBank mRNA | BC168698.1 | - | 15, 16 | 1816 | 7938988 | |