Search
Search

Sema3f| Gene Symbol: Sema3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108185.1 | NP_001101655.1 | 7, 8 | 844 | ||
| XM_006243778.2 | XP_006243840.1 | 7, 8 | 1399 | |||
| XM_008766532.1 | XP_008764754.1 | 7, 8 | 727 | |||
| XM_008766533.1 | XP_008764755.1 | 4, 5 | 1688 | |||